Assay Design



Gene Transcripts

Gene Symbol
Pcdha5
Entrez Gene ID
393087
Gene Name
protocadherin alpha 5
Gene Aliases
rCNRv05
Chromosome Location
Chr.18: 29980268 - 30215897 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199505.4 NP_955799.1 1 - 1 2202 84
GenBank mRNA AB113388.1 - 1 - 1 2020 84
AY573979.1 - 1 - 1 2202 84

View Full Product Details