Assay Design



Gene Transcripts

Gene Symbol
Ammecr1l
Entrez Gene ID
307526
Gene Name
AMME chromosomal region gene 1-like
Gene Aliases
RGD1307915
Chromosome Location
Chr.18: 24539432 - 24563576 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254593.2 XP_006254655.1 1 - 2 367 138

View Full Product Details