Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107399.1 | NP_001100869.1 | 4 - 5 | 787 | 101 |
XM_006254590.2 | XP_006254652.1 | 4 - 5 | 1601 | 101 | |
XM_006254591.2 | XP_006254653.1 | 4 - 5 | 1638 | 101 | |
XM_006254593.2 | XP_006254655.1 | 4 - 5 | 1038 | 101 | |
XM_008772042.1 | XP_008770264.1 | 4 - 5 | 1638 | 101 |