Assay Design



Gene Transcripts

Gene Symbol
Pcdhb12
Entrez Gene ID
25133
Gene Name
protocadherin beta 12
Gene Aliases
Pcdh3
Chromosome Location
Chr.18: 30495596 - 30518524 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254627.2 XP_006254689.1 2 - 2 1770 121

View Full Product Details