Assay Design



Gene Transcripts

Gene Symbol
Pcdhb19
Entrez Gene ID
548106
Gene Name
protocadherin beta 19
Gene Aliases
-
Chromosome Location
Chr.18: 30572252 - 30577286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001056051.4 XP_001056051.1 1 - 1 4978 83

View Full Product Details