Assay Design



Gene Transcripts

Gene Symbol
RGD735029
Entrez Gene ID
307480
Gene Name
SEL1 domain containing protein RGD735029
Gene Aliases
Dele
Chromosome Location
Chr.18: 31396898 - 31409914 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199493.1 NP_955787.1 1 - 2 67 87
GenBank mRNA BC064660.1 - 1 - 2 67 87
FQ211991.1 - - 123 87

View Full Product Details