Assay Design



Gene Transcripts

Gene Symbol
RGD735029
Entrez Gene ID
307480
Gene Name
SEL1 domain containing protein RGD735029
Gene Aliases
Dele
Chromosome Location
Chr.18: 31396898 - 31409914 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199493.1 NP_955787.1 2 - 3 185 87
XM_006254634.2 XP_006254696.1 2 - 3 292 87
GenBank mRNA BC064660.1 - 2 - 3 185 87
FQ211991.1 - - 241 87

View Full Product Details