Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014012.2 | NP_001014034.2 | 4 - 5 | 517 | 90 |
XM_006254575.2 | XP_006254637.1 | 3 - 4 | 427 | 90 | |
GenBank mRNA | CK481952.1 | - | 4 - 5 | 533 | 90 |
CV075096.1 | - | 1 - 2 | 120 | 90 |