Assay Design



Gene Transcripts

Gene Symbol
Sh3rf2
Entrez Gene ID
307472
Gene Name
SH3 domain containing ring finger 2
Gene Aliases
Posh2, Ppp1r39
Chromosome Location
Chr.18: 36371041 - 36471020 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034187.1 NP_001029359.1 4 - 5 1213 93
GenBank mRNA BC100155.1 - 4 - 5 1213 93

View Full Product Details