Assay Design



Gene Transcripts

Gene Symbol
Pcdhb20
Entrez Gene ID
680203
Gene Name
protocadherin beta 20
Gene Aliases
Pcdhb14
Chromosome Location
Chr.18: 30581530 - 30584905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109395.1 NP_001102865.1 1 - 1 3081 105

View Full Product Details