Assay Design



Gene Transcripts

Gene Symbol
Wdr55
Entrez Gene ID
307494
Gene Name
WD repeat domain 55
Gene Aliases
LRRG00133, RGD1305640
Chromosome Location
Chr.18: 29605217 - 29608730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017932.1 NP_001017932.1 4 - 5 646 111
GenBank mRNA BC127460.1 - 4 - 5 601 111

View Full Product Details