Assay Design



Gene Transcripts

Gene Symbol
Cbr3
Entrez Gene ID
304078
Gene Name
carbonyl reductase 3
Gene Aliases
-
Chromosome Location
Chr.11: 33909417 - 33917674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107110.1 NP_001100580.1 2 - 3 508 59
GenBank mRNA BC166553.1 - 2 - 3 469 59
FQ234106.1 - 2 - 3 486 59

View Full Product Details