Assay Design



Gene Transcripts

Gene Symbol
Psmg1
Entrez Gene ID
288236
Gene Name
proteasome (prosome, macropain) assembly chaperone 1
Gene Aliases
Dscr2
Chromosome Location
Chr.11: 36327798 - 36337281 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105891.1 NP_001099361.1 4 - 5 826 73
XM_006248154.2 XP_006248216.1 5 - 6 711 73
XM_008768571.1 XP_008766793.1 5 - 6 983 73

View Full Product Details