Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191992.1 | NP_001178921.1 | 3 - 4 | 415 | 76 |
XM_006242237.1 | XP_006242299.1 | 3 - 4 | 524 | 76 | |
XM_006242238.2 | XP_006242300.1 | 4 - 5 | 1058 | 76 | |
XM_008765776.1 | XP_008763998.1 | 3 - 4 | 798 | 76 |