Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017177.1 | NP_058873.1 | 7 - 8 | 1092 | 94 |
XM_006242181.2 | XP_006242243.2 | 7 - 8 | 1758 | 94 | |
XM_008765677.1 | XP_008763899.1 | 7 - 8 | 1755 | 94 | |
GenBank mRNA | AB006607.1 | - | 7 - 8 | 1092 | 94 |
BC060515.1 | - | 7 - 8 | 1068 | 94 |