Assay Design



Gene Transcripts

Gene Symbol
Zcrb1
Entrez Gene ID
362990
Gene Name
zinc finger CCHC-type and RNA binding motif 1
Gene Aliases
RGD1309851
Chromosome Location
Chr.7: 134588902 - 134602239 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034940.1 NP_001030112.1 5 - 6 473 100
GenBank mRNA BC099747.1 - 5 - 6 473 100

View Full Product Details