Assay Design



Gene Transcripts

Gene Symbol
Dos
Entrez Gene ID
314622
Gene Name
downstream of Stk11
Gene Aliases
-
Chromosome Location
Chr.7: 12433422 - 12441048 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108070.1 NP_001101540.1 3 - 4 684 100

View Full Product Details