Assay Design
Gene Transcripts
- Gene Symbol
- Palm
- Entrez Gene ID
-
170673
- Gene Name
- paralemmin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 12708024 - 12721055 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC072525.1 |
- |
- |
369 |
86 |
View Full Product Details