Assay Design
Gene Transcripts
- Gene Symbol
- Cyp4f4
- Entrez Gene ID
-
286904
- Gene Name
- cytochrome P450, family 4, subfamily f, polypeptide 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 14891062 - 14907250 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173123.1 |
NP_775146.1 |
4 - 5 |
534 |
83 |
GenBank mRNA |
U39206.1 |
- |
4 - 5 |
534 |
83 |
View Full Product Details