Assay Design



Gene Transcripts

Gene Symbol
Prss57
Entrez Gene ID
408241
Gene Name
protease, serine, 57
Gene Aliases
Df2, Prssl1
Chromosome Location
Chr.7: 12798725 - 12804668 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003956.1 NP_001003956.1 3 - 4 384 100
XM_006240979.2 XP_006241041.1 3 - 4 527 100
XM_008765175.1 XP_008763397.1 3 - 4 527 100

View Full Product Details