Assay Design
Gene Transcripts
- Gene Symbol
- Rps15
- Entrez Gene ID
-
29285
- Gene Name
- ribosomal protein S15
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 12282134 - 12283573 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AA892895.1 |
- |
3 - 4 |
|
76 |
View Full Product Details