Assay Design



Gene Transcripts

Gene Symbol
En1
Entrez Gene ID
685360
Gene Name
engrailed homeobox 1
Gene Aliases
-
Chromosome Location
Chr.13: 36532758 - 36537888 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001056699.6 XP_001056699.3 1 - 2 918 146

View Full Product Details