Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271109.1 | NP_001258038.1 | 11 - 12 | 1450 | 73 |
NM_001271110.1 | NP_001258039.1 | 11 - 12 | 1478 | 73 | |
NM_080479.2 | NP_536727.2 | 11 - 12 | 1450 | 73 | |
XM_006237924.2 | XP_006237986.1 | 11 - 12 | 1540 | 73 | |
GenBank mRNA | AY724507.1 | - | 11 - 12 | 1463 | 73 |
BC086998.1 | - | 9 - 10 | 2407 | 73 | |
BC107467.1 | - | 11 - 12 | 1450 | 73 | |
FQ229095.1 | - | 11 - 12 | 1463 | 73 |