Assay Design



Gene Transcripts

Gene Symbol
Lce1m
Entrez Gene ID
688413
Gene Name
late cornified envelope 1M
Gene Aliases
-
Chromosome Location
Chr.2: 193333800 - 193335002 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109500.1 NP_001102970.1 1 - 2 44 67

View Full Product Details