Assay Design



Gene Transcripts

Gene Symbol
Ghrhr
Entrez Gene ID
25321
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRHREC
Chromosome Location
Chr.4: 85587321 - 85601666 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012850.1 NP_036982.1 8 - 9 842 80
XM_008762872.1 XP_008761094.1 8 - 9 2205 80
GenBank mRNA AF122055.1 - 8 - 9 815 80
L01407.1 - 8 - 9 842 80

View Full Product Details