Assay Design



Gene Transcripts

Gene Symbol
Scrn1
Entrez Gene ID
502776
Gene Name
secernin 1
Gene Aliases
-
Chromosome Location
Chr.4: 84678978 - 84740988 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025063.1 NP_001020234.1 5 - 6 915 84
XM_006236512.2 XP_006236574.1 5 - 6 903 84
XM_008762916.1 XP_008761138.1 5 - 6 767 84
GenBank mRNA BC079152.1 - 5 - 6 915 84

View Full Product Details