Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013031.1 | NP_037163.1 | 5 - 6 | 708 | 104 |
XM_006231664.2 | XP_006231726.1 | 5 - 6 | 1549 | 104 | |
XM_008760515.1 | XP_008758737.1 | 4 - 5 | 908 | 104 | |
XM_008760516.1 | XP_008758738.1 | 5 - 6 | 1554 | 104 | |
XM_008760517.1 | XP_008758739.1 | 5 - 6 | 1549 | 104 | |
XM_008760518.1 | XP_008758740.1 | 4 - 5 | 908 | 104 | |
GenBank mRNA | L00603.1 | - | 5 - 6 | 708 | 104 |
M97381.1 | - | 5 - 6 | 679 | 104 |