Assay Design



Gene Transcripts

Gene Symbol
Habp2
Entrez Gene ID
292126
Gene Name
hyaluronan binding protein 2
Gene Aliases
Fsap, Habp, Phbp
Chromosome Location
Chr.1: 277068713 - 277104567 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001505.1 NP_001001505.1 11 - 12 1394 93
XM_006231631.2 XP_006231693.1 11 - 12 1442 93
XM_006231632.2 XP_006231694.1 10 - 11 1331 93
XM_006231633.2 XP_006231695.1 10 - 11 1328 93
GenBank mRNA AB177406.1 - 11 - 12 1394 93
BC129080.1 - 10 - 11 1280 93

View Full Product Details