Assay Design



Gene Transcripts

Gene Symbol
Hlx
Entrez Gene ID
364069
Gene Name
H2.0-like homeobox
Gene Aliases
Hlx1
Chromosome Location
Chr.13: 102637967 - 102643376 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077674.1 NP_001071142.1 1 - 2 912 77
GenBank mRNA BC127513.1 - 1 - 2 912 77

View Full Product Details