Assay Design



Gene Transcripts

Gene Symbol
Usf1
Entrez Gene ID
83586
Gene Name
upstream transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.13: 89797750 - 89805561 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031777.2 NP_113965.1 2 - 3 142 105
XM_006250266.2 XP_006250328.1 2 - 3 327 105
GenBank mRNA BC088849.1 - 2 - 3 142 105

View Full Product Details