Assay Design



Gene Transcripts

Gene Symbol
Casq1
Entrez Gene ID
686019
Gene Name
calsequestrin 1 (fast-twitch, skeletal muscle)
Gene Aliases
-
Chromosome Location
Chr.13: 90592676 - 90602365 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159594.1 NP_001153066.1 8 - 9 1039 118
GenBank mRNA FM056035.1 - 5 - 6 344 118
FQ216689.1 - 8 - 9 616 118
FQ224875.1 - 9 - 10 866 118

View Full Product Details