Assay Design
Gene Transcripts
- Gene Symbol
- Hhipl2
- Entrez Gene ID
-
317051
- Gene Name
- hedgehog interacting protein-like 2
- Gene Aliases
-
RGD1311317
- Chromosome Location
-
Chr.13: 101814574 - 101834641 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_229754.7 |
XP_229754.4 |
8 - 9 |
1881 |
71 |
View Full Product Details