Assay Design
Gene Transcripts
- Gene Symbol
- Tp53bp2
- Entrez Gene ID
-
305025
- Gene Name
- tumor protein p53 binding protein, 2
- Gene Aliases
-
Trp53bp2
- Chromosome Location
-
Chr.13: 100817204 - 100873837 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_223012.8 |
XP_223012.6 |
9 - 10 |
1465 |
84 |
View Full Product Details