Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014787.1 | NP_001014787.1 | 5 - 6 | 1381 | 140 |
XM_006250286.2 | XP_006250348.1 | 5 - 6 | 1540 | 140 | |
XM_006250288.2 | XP_006250350.1 | 4 - 5 | 1482 | 140 | |
XM_006250289.2 | XP_006250351.1 | 4 - 5 | 1319 | 140 | |
GenBank mRNA | BC092192.1 | - | 5 - 6 | 1381 | 140 |