Assay Design



Gene Transcripts

Gene Symbol
Traf3ip3
Entrez Gene ID
360900
Gene Name
TRAF3 interacting protein 3
Gene Aliases
RGD1304848
Chromosome Location
Chr.13: 111892744 - 111917368 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014132.1 NP_001014154.1 14 - 15 1546 108
XM_008769865.1 XP_008768087.1 13 - 14 2211 108
GenBank mRNA BC089843.1 - 14 - 15 1546 108

View Full Product Details