Assay Design



Gene Transcripts

Gene Symbol
Zkscan5
Entrez Gene ID
304275
Gene Name
zinc finger with KRAB and SCAN domains 5
Gene Aliases
Zfp95
Chromosome Location
Chr.12: 11154807 - 11176002 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107119.1 NP_001100589.1 5 - 6 2056 102

View Full Product Details