Assay Design



Gene Transcripts

Gene Symbol
Asmtl
Entrez Gene ID
288527
Gene Name
acetylserotonin O-methyltransferase-like
Gene Aliases
-
Chromosome Location
Chr.12: 18532090 - 18534761 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105915.2 NP_001099385.2 4 - 5 427 72
XM_006249001.2 XP_006249063.1 5 - 6 544 72
GenBank mRNA FM053536.1 - 2 - 3 96 72

View Full Product Details