Assay Design



Gene Transcripts

Gene Symbol
Gabra2
Entrez Gene ID
289606
Gene Name
gamma-aminobutyric acid (GABA) A receptor, alpha 2
Gene Aliases
-
Chromosome Location
Chr.14: 39662377 - 39799328 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135779.1 NP_001129251.1 6 - 7 594 130
XM_006250931.2 XP_006250993.1 6 - 7 658 130

View Full Product Details