Assay Design



Gene Transcripts

Gene Symbol
Ccng2
Entrez Gene ID
29157
Gene Name
cyclin G2
Gene Aliases
-
Chromosome Location
Chr.14: 16267572 - 16276068 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105725.2 NP_001099195.1 2 - 3 322 152
GenBank mRNA BC169085.1 - 2 - 3 322 152

View Full Product Details