Assay Design



Gene Transcripts

Gene Symbol
Amtn
Entrez Gene ID
689404
Gene Name
amelotin
Gene Aliases
-
Chromosome Location
Chr.14: 21286509 - 21300620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044296.1 NP_001037761.1 1 - 2 77 96
GenBank mRNA DQ198381.1 - 1 - 2 77 96

View Full Product Details