Assay Design



Gene Transcripts

Gene Symbol
Fam47e
Entrez Gene ID
686140
Gene Name
family with sequence similarity 47, member E
Gene Aliases
Fam47e-ps1
Chromosome Location
Chr.14: 16991496 - 17027568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770095.1 XP_008768317.1 8 - 9 1293 107
XM_008770096.1 XP_008768318.1 7 - 8 1200 107

View Full Product Details