Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100509.1 | NP_001093979.1 | 5 - 6 | 386 | 126 |
GenBank mRNA | BC107950.1 | - | 5 - 6 | 307 | 126 |
DY570284.1 | - | 5 - 6 | 386 | 126 | |
FQ213949.1 | - | 5 - 6 | 334 | 126 | |
FQ214043.1 | - | 5 - 6 | 339 | 126 | |
FQ227742.1 | - | 5 - 6 | 343 | 126 |