Assay Design



Gene Transcripts

Gene Symbol
Cox20
Entrez Gene ID
289278
Gene Name
COX20 cytochrome C oxidase assembly factor
Gene Aliases
Fam36a, RGD1309105
Chromosome Location
Chr.13: 96219853 - 96224023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105976.1 NP_001099446.1 1 - 2 295 135
GenBank mRNA FQ229252.1 - 1 - 2 57 135

View Full Product Details