Assay Design



Gene Transcripts

Gene Symbol
Angel2
Entrez Gene ID
305035
Gene Name
angel homolog 2 (Drosophila)
Gene Aliases
RGD1305056
Chromosome Location
Chr.13: 109504430 - 109520971 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135119.2 NP_001128591.2 5 - 6 1203 150
XM_006250462.2 XP_006250524.1 6 - 7 1208 150
XM_008769852.1 XP_008768074.1 5 - 6 1205 150

View Full Product Details