Assay Design
Gene Transcripts
- Gene Symbol
- Cst8
- Entrez Gene ID
-
29679
- Gene Name
- cystatin 8 (cystatin-related epididymal specific)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 143129281 - 143135918 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_019258.1 |
NP_062131.1 |
3 - 4 |
542 |
128 |
GenBank mRNA |
AF090692.1 |
- |
3 - 4 |
542 |
128 |
View Full Product Details