Assay Design



Gene Transcripts

Gene Symbol
Otor
Entrez Gene ID
366206
Gene Name
otoraplin
Gene Aliases
-
Chromosome Location
Chr.3: 137154086 - 137157341 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108960.1 NP_001102430.1 1 - 2 129 105

View Full Product Details