Assay Design



Gene Transcripts

Gene Symbol
Syndig1
Entrez Gene ID
362235
Gene Name
synapse differentiation inducing 1
Gene Aliases
DSPC2, RGD1310753, Tmem90b
Chromosome Location
Chr.3: 145032177 - 145199196 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025020.1 NP_001020191.1 3 - 4 721 67
GenBank mRNA BC092131.1 - 3 - 4 721 67

View Full Product Details