Assay Design
Gene Transcripts
- Gene Symbol
- Gid8
- Entrez Gene ID
-
296466
- Gene Name
- GID complex subunit 8
- Gene Aliases
-
Bwk1, TWA1
- Chromosome Location
-
Chr.3: 176217128 - 176223645 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198743.1 |
NP_942038.1 |
3 - 4 |
384 |
70 |
GenBank mRNA |
AY156921.1 |
- |
3 - 4 |
384 |
70 |
View Full Product Details