Assay Design
Gene Transcripts
- Gene Symbol
- Tmem27
- Entrez Gene ID
-
57395
- Gene Name
- transmembrane protein 27
- Gene Aliases
-
NX-17, Nx17, collectrin
- Chromosome Location
-
Chr.X: 32118082 - 32153687 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC078838.1 |
- |
4 - 5 |
380 |
114 |
View Full Product Details