Assay Design



Gene Transcripts

Gene Symbol
Tmem27
Entrez Gene ID
57395
Gene Name
transmembrane protein 27
Gene Aliases
NX-17, Nx17, collectrin
Chromosome Location
Chr.X: 32118082 - 32153687 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020976.1 NP_066125.1 5 - 6 521 117
GenBank mRNA AF178086.1 - 5 - 6 521 117
BC078838.1 - 5 - 6 578 117

View Full Product Details